Thermo Scientific Chemicals

Nimodipine, 98+%, Thermo Scientific Chemicals

Catalog number: J61287.ME
500 mg, Each
Thermo Scientific Chemicals

Nimodipine, 98+%, Thermo Scientific Chemicals

Catalog number: J61287.ME
500 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61287.ME
also known as J61287-ME
Price (USD)
Price: 101.00
Online price: 87.65
Your price:
Quantity
-

Chemical Identifiers

CAS
66085-59-4
IUPAC Name
3-(2-methoxyethyl) 5-propan-2-yl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
Molecular Formula
C21H26N2O7
InChI Key
UIAGMCDKSXEBJQ-UHFFFAOYNA-N
SMILES
COCCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC(=C1)[N+]([O-])=O)C(=O)OC(C)C
Form
Powder
Purity
≥98.0%

Description

Potent L-type calcium channel antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text