Sodium acetate, 3M aq. soln., pH 4.5, autoclaved
Sodium acetate, 3M aq. soln., pH 4.5, autoclaved
Sodium acetate, 3M aq. soln., pH 4.5, autoclaved
Sodium acetate, 3M aq. soln., pH 4.5, autoclaved
Thermo Scientific Chemicals

Sodium acetate, 3M aq. soln., pH 4.5, autoclaved

CAS: 127-09-3 | C2H3NaO2 | 82.03 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
125 mL
250 mL
產品號碼 J61288.AK
亦稱為 J61288-AK
價格 (TWD)
-
申請報價
Quantity:
250 mL
Request bulk or custom format
規格
CAS127-09-3
ChEBICHEBI:32954
Chemical Name or MaterialSodium acetate
Concentration3M aq. soln.
Formula Weight82.03
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only