Dobutamine hydrochloride
Dobutamine hydrochloride
Dobutamine hydrochloride
Thermo Scientific Chemicals

Dobutamine hydrochloride

CAS: 49745-95-1 | C18H24ClNO3 | 337.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61333.MD
also known as J61333-MD
Price (EUR)
-
Quantity:
250 mg
Request bulk or custom format
Chemical Identifiers
CAS49745-95-1
IUPAC Namehydrogen 4-(2-{[4-(4-hydroxyphenyl)butan-2-yl]amino}ethyl)benzene-1,2-diol chloride
Molecular FormulaC18H24ClNO3
InChI KeyBQKADKWNRWCIJL-UHFFFAOYNA-N
SMILES[H+].[Cl-].CC(CCC1=CC=C(O)C=C1)NCCC1=CC=C(O)C(O)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (HPLC)≥98.0%
Appearance (Color)White to off-white
Dobutamine hydrochloride is a catecholamine that acts as a β-adrenergic receptor agonist. It has its strongest effects on β1 receptors, with lesser but significant β2 receptor activation and only weak α1 receptor actions. As a result, it strongly increases cardiac contractility with modest chronotropic, arrhythmogenic, and vascular side effects. It is also a polyphenolic antioxidant and can inhibit all human carbonic anhydrase isoforms with Ki values near 1M.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dobutamine hydrochloride is a catecholamine that acts as a β-adrenergic receptor agonist. It has its strongest effects on β1 receptors, with lesser but significant β2 receptor activation and only weak α1 receptor actions. As a result, it strongly increases cardiac contractility with modest chronotropic, arrhythmogenic, and vascular side effects. It is also a polyphenolic antioxidant and can inhibit all human carbonic anhydrase isoforms with Ki values near 1 μM.

Solubility
Soluble in water or ethanol with warming. Also soluble in methanol

Notes
Solutions oxidize readily at pH greater than 11. Protect solutions from light
RUO – Research Use Only

General References:

  1. R.R.Tuttle; J.Mills. Dobutamine: development of a new catecholamine to selectively increase cardiac contractility. Circulation Research. 1975, 36,(1), 185-196.
  2. Y.Huang; K.H.Kwok; N.W.Chan; C.W.Lau; Z.Y.Chen. Role of endothelium and K+ channels in dobutamine-induced relaxation in rat mesenteric artery. Clinical and Experimental Pharmacology and Physiology. 1998, 25,(6), 405-411.