Sodium phosphate, 0.2M buffer soln., pH 8.5
Thermo Scientific Chemicals
Sodium phosphate, 0.2M buffer soln., pH 8.5
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number J61372.K2
also known as J61372-K2
Price (USD)/ Each
118.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
139.00 
Save 20.35 (15%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
118.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
139.00 
Save 20.35 (15%)
Add to cart
Sodium phosphate, 0.2M buffer soln., pH 8.5
Catalog numberJ61372.K2
Price (USD)/ Each
118.65
Special Offer
Online exclusive
Ends: 15-Oct-2024
139.00 
Save 20.35 (15%)
-
Add to cart
Sodium phosphate buffer is used routinely in the life sciences. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium phosphate buffer is used routinely in the life sciences. It is been also used for PEGylation of proteins at pH 7.4 and a concentration of 50 mM. Sodium phosphate buffer solution may be used in the preparation of a wide variety of phosphate based biological buffers for use in cell and molecular biology application.

Solubility
Soluble in water (121 mg/ml at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong acids.
RUO – Research Use Only

General References:

  1. CR Shaw.; R Prasad. Starch gel electrophoresis of enzymes—a compilation of recipes. Biochemical genetics. 1970, 4,(2), 297-320.
  2. AM Maxam.; W Gilbert. A new method for sequencing DNA. PNAS. 1977, 74,(2), 560-564.