Tulobuterol Hydrochloride, 98%
Tulobuterol Hydrochloride, 98%
Tulobuterol Hydrochloride, 98%
Thermo Scientific Chemicals

Tulobuterol Hydrochloride, 98%

beta-2-adrenergic receptor agonist | CAS: 56776-01-3 | C12H18ClNO·HCl
Have Questions?
Catalog NumberQuantity
J61448.03
also known as J61448-03
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J61448.03
also known as J61448-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Assay98%
CAS56776-01-3
DescriptionWhite
For Use With (Application)beta-2-adrenergic receptor agonist
FormPowder
View more
beta-2-adrenergic receptor agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
2°C to 8°C