EPPS, 0.2M buffer soln., pH 8.5
EPPS, 0.2M buffer soln., pH 8.5
EPPS, 0.2M buffer soln., pH 8.5
Thermo Scientific Chemicals

EPPS, 0.2M buffer soln., pH 8.5

CAS: 16052-06-5 | C9H20N2O4S | 252.329 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 mL
250 mL
產品號碼 J61476.AE
亦稱為 J61476-AE
價格 (TWD)
-
Quantity:
100 mL
Request bulk or custom format
規格
CAS16052-06-5
ChEBICHEBI:42298
Chemical Name or MaterialEPPS buffer soln.
IUPAC Name3-[4-(2-hydroxyethyl)piperazin-1-yl]propane-1-sulfonic acid
InChI KeyOWXMKDGYPWMGEB-UHFFFAOYSA-N
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only