Bradford Dye Reagent, Ready-to-Use soln.
Thermo Scientific Chemicals

Bradford Dye Reagent, Ready-to-Use soln.

Suitable for protein concentration determination (100 to 1500μg)
Have Questions?
Catalog NumberQuantity
J61522.K2
also known as J61522-K2
1 L
Catalog number J61522.K2
also known as J61522-K2
Price (TWD)
-
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialBradford Dye Reagent
ColorBlue
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: PHOSPHORIC ACID SOLUTION
Health Hazard 1H290-H302+H332-H314-H335-H371
Health Hazard 2GHS H Statement
H314-H318-H371
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause damage to organs.
View more
The Bradford Reagent is suitable for protein concentration determination (100-1500μg).Bradford dye reagent is used in the determination of protein content in cell fractions and assessing the protein concentrations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Frequently asked questions (FAQs)

At what temperature should I store Bradford Dye Reagent, Ready-to-Use soln.?

The Bradford Dye Reagent, Ready-to-Use soln. should be stored refrigerated at 2-8 degrees C.