TRIS, 1.0M buffer soln., pH 7.8
TRIS, 1.0M buffer soln., pH 7.8
TRIS, 1.0M buffer soln., pH 7.8
Thermo Scientific Chemicals

TRIS, 1.0M buffer soln., pH 7.8

CAS: 77-86-1 | C4H11NO3 | 121.136 g/mol
Have Questions?
Catalog NumberQuantity
J61579.K2
also known as J61579-K2
1 L
Catalog number J61579.K2
also known as J61579-K2
Price (TWD)
-
Quantity:
1 L
Request bulk or custom format
Specifications
CAS77-86-1
ChEBICHEBI:9754
Chemical Name or MaterialTRIS buffer soln.
IUPAC Name2-amino-2-(hydroxymethyl)propane-1,3-diol
InChI KeyLENZDBCJOHFCAS-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only