L-Tyrosine disodium salt dihydrate
L-Tyrosine disodium salt dihydrate
L-Tyrosine disodium salt dihydrate
Thermo Scientific Chemicals

L-Tyrosine disodium salt dihydrate

CAS: 122666-87-9 | C9H13NNa2O5 | 261.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Catalog number J61770.22
also known as J61770-22
Price (EUR)
99,50
Each
Quantity:
100 g
Request bulk or custom format
Price (EUR)
99,50
Each
Chemical Identifiers
CAS122666-87-9
IUPAC Namedisodium (2S)-2-amino-3-(4-oxidophenyl)propanoate dihydrate
Molecular FormulaC9H13NNa2O5
InChI KeyWFTCFVUQORELJZ-CMIWSEPMNA-L
SMILESO.O.[Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
FormPowder
Assay (special)Endotoxin: ≤0.04 EU/mg
Heavy Metals (as Pb)≤15 ppm (≤0.0015%)
pH10.5 to 12.0
View more
Tyrosine is a polar, naturally occurring, non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tyrosine is a polar, naturally occurring, non-essential amino acid. It is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies.

Solubility
Soluble in water.

Notes
Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only