Thermo Scientific Chemicals

DL-Isoproterenol hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: J61788.14
25 g, Each
Thermo Scientific Chemicals

DL-Isoproterenol hydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: J61788.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61788.14
also known as J61788-14
Price (USD)
Quantity
-

Description

Isoproterenol hydrochloride is used as a selective β-adrenergic agonist, increases cytosolic cAMP.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isoproterenol hydrochloride is used as a selective β-adrenergic agonist, increases cytosolic cAMP.

Solubility
Soluble in water (50 mg/ml at 25°C), ethanol (20 mg/ml), methanol, DMSO (50 mg/ml at 25°C), and phosphate buffered saline (50 mM).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text