Ammonium peroxydisulfate, Electrophoresis Grade
Ammonium peroxydisulfate, Electrophoresis Grade
Ammonium peroxydisulfate, Electrophoresis Grade
Ammonium peroxydisulfate, Electrophoresis Grade
Thermo Scientific Chemicals

Ammonium peroxydisulfate, Electrophoresis Grade

CAS: 7727-54-0 | H8N2O8S2 | 228.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number J61856.14
also known as J61856-14
Price (EUR)
33,20
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
33,20
Each
Add to cart
Chemical Identifiers
CAS7727-54-0
IUPAC Namediammonium [(sulfonatoperoxy)sulfonyl]oxidanide
Molecular FormulaH8N2O8S2
InChI KeyROOXNKNUYICQNP-UHFFFAOYSA-N
SMILES[NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥98.0%
Heavy Metals (as Pb)≤0.001% (≤10ppm)
FormCrystals or powder or crystalline powder
CommentDNase, RNase and Protease not detected
Free acid (titration)≤0.04meq/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only