MOPS, 1.0M buffer soln., pH 8.0
MOPS, 1.0M buffer soln., pH 8.0
MOPS, 1.0M buffer soln., pH 8.0
MOPS, 1.0M buffer soln., pH 8.0
Thermo Scientific Chemicals

MOPS, 1.0M buffer soln., pH 8.0

CAS: 1132-61-2 | C7H15NO4S | 209.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J61958.AK
also known as J61958-AK
Price (TWD)
-
Request A Quote
Quantity:
250 mL
Request bulk or custom format
Specifications
CAS1132-61-2
ChEBICHEBI:44115
Chemical Name or MaterialMOPS buffer soln.
ColorColorless
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only