Lansoprazole, 98+%
Lansoprazole, 98+%
Lansoprazole, 98+%
Thermo Scientific Chemicals

Lansoprazole, 98+%

A proton pump inhibitor | CAS: 103577-45-3 | C16H14F3N3O2S | 369.362 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62008.ME
also known as J62008-ME
Price (TWD)
-
Quantity:
500 mg
Request bulk or custom format
Specifications
CAS103577-45-3
ChEBICHEBI:6375
Chemical Name or MaterialLansoprazole
ColorWhite
Formula Weight369.36
View more
A proton pump inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only