Thermo Scientific™

Water, DEPC-Treated, Thermo Scientific™

1L, Each
Thermo Scientific™

Water, DEPC-Treated, Thermo Scientific™

1L, Each
Quantity
Catalog number: J62087.K2
also known as J62087-K2
Price (USD)
102.00 / Each
Your Price:
Quantity
-

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used as mobile phase solvent in LC-MS analysis.

Solubility
Soluble in water. Insoluble in polar liquids.

Notes
Suitable for any general chemical storage area. Protect from freezing. Incompatib
RUO – Research Use Only

Figures