1-(1-Naphthyl)piperazine hydrochloride, Thermo Scientific Chemicals
1-(1-Naphthyl)piperazine hydrochloride, Thermo Scientific Chemicals
1-(1-Naphthyl)piperazine hydrochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(1-Naphthyl)piperazine hydrochloride, Thermo Scientific Chemicals

Have Questions?
產品號碼Quantity
J62097.03
亦稱為 J62097-03
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 J62097.03
亦稱為 J62097-03
價格 (TWD)
-
Quantity:
1 g
Request bulk or custom format
化學識別
CAS104113-71-5
IUPAC Name1-(naphthalen-1-yl)piperazine hydrochloride
Molecular FormulaC14H17ClN2
InChI KeyZYVYPNZFOCZLEM-UHFFFAOYSA-N
SMILESCl.C1CN(CCN1)C1=C2C=CC=CC2=CC=C1
檢視更多
規格Specification Sheet規格表
Assay (HPLC)≥98.0%
1H-NMR (Chloroform)Conforms to structure
FormCrystalline solid
5-HT1 serotonin receptor agonist. 5-HT2 serotonin receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • 5-HT1 serotonin receptor agonist
  • 5-HT2 serotonin receptor antagonist
RUO – Research Use Only