Borate, 0.5M buffer soln., pH 9.0
Borate, 0.5M buffer soln., pH 9.0
Borate, 0.5M buffer soln., pH 9.0
Thermo Scientific Chemicals

Borate, 0.5M buffer soln., pH 9.0

CAS: 1330-43-4 | B4Na2O7 | 201.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
500 mL
Catalog number J62125.AK
also known as J62125-AK
Price (EUR)
72,60
Each
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
72,60
Each
Specifications
CAS1330-43-4
Chemical Name or MaterialBorate buffer soln.
Health Hazard 1H319-H360FD
Health Hazard 2GHS H Statement
H360
May damage fertility or the unborn child.
Health Hazard 3P201-P202-P264b-P280i-P281-P305+P351+P338-P308+P313-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only