Fluoresceinamine, isomer I, 96%
Fluoresceinamine, isomer I, 96%
Fluoresceinamine, isomer I, 96%
Fluoresceinamine, isomer I, 96%
Thermo Scientific Chemicals

Fluoresceinamine, isomer I, 96%

A fluorescent labeling reagent | CAS: 3326-34-9 | C20H13NO5 | 347.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number J62175.03
also known as J62175-03
Price (EUR)
132,00
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (EUR)
132,00
Each
Add to cart
Specifications
Beilstein48395
CAS3326-34-9
Chemical Name or MaterialFluoresceinamine
EINECS Number222-043-6
Formula Weight347.32
View more
5-aminofluorescein is also used to prepare FITC Isomer I (CDX-F0011, CDX-F0020). It is a chemical used as molecular probe and important as fluorescent dye and for derivatives. Fluorescent labeling reagent for proteins. Used in the fluorescent antibody technique for rapid identification of pathogens.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-aminofluorescein is also used to prepare FITC Isomer I (CDX-F0011, CDX-F0020). It is a chemical used as molecular probe and important as fluorescent dye and for derivatives. Fluorescent labeling reagent for proteins. Used in the fluorescent antibody technique for rapid identification of pathogens.

Solubility
Soluble in water (partly), methanol (10 mg/ml), and acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Light and Moisture Sensitive
RUO – Research Use Only

General References:

  1. KS Bronk.; KL Michael.; P Pantano.; DR Walt. Combined imaging and chemical sensing using a single optical imaging fiber. Analytical chemistry.1995, 67, (17), 2750-2757.
  2. HD Duong.; OJ Sohn.; HT Lam.; JI Rhee. An optical pH sensor with extended detection range based on fluoresceinamine covalently bound to sol-gel support. Microchemical journal.2006, 84, (1-2), 50-55.