Neostigmine bromide
Neostigmine bromide
Neostigmine bromide
Thermo Scientific Chemicals

Neostigmine bromide

A reversible acetylcholinesterase inhibitor | CAS: 114-80-7 | C12H19BrN2O2 | 303.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62300.03
also known as J62300-03
Price (EUR)
71,90
Each
Quantity:
1 g
Request bulk or custom format
Price (EUR)
71,90
Each
Chemical Identifiers
CAS114-80-7
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
FormPowder or crystals or crystalline powder
Assay from Suppliers CofA≥97.5%
Melting Point171.0-177.0°C
A reversible acetylcholinesterase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only