Thermo Scientific Chemicals

Magnesium chloride, 1M aq. soln., Thermo Scientific Chemicals

Catalog number: J62411.AE
100 mL, Each
Thermo Scientific Chemicals

Magnesium chloride, 1M aq. soln., Thermo Scientific Chemicals

Catalog number: J62411.AE
100 mL, Each
Quantity
Catalog number: J62411.AE
also known as J62411-AE
Price (USD)
Price: 86.10
Online price: 73.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7786-30-3
IUPAC Name
magnesium(2+) dichloride
Molecular Formula
Cl2Mg
InChI Key
TWRXJAOTZQYOKJ-UHFFFAOYSA-L
SMILES
[Mg++].[Cl-].[Cl-]
Transition Temperature
Component is dissolve in 18.2 megahom, UV treated water and filtered through sub-micron filter
Appearance (Color)
Clear and colorless solution

Description

Magnesium chloride, 1M aq. soln., sterile is used as an excellent magnesium ion source for synthesis and biology research. Magnesium chloride is widely used to supply the magnesium ion in various molecular biology applications, including PCR reactions and buffers. It was used to prepare buffer to lyse cells for immunoprecipitation1 and to prepare serum-free medium for electrophysiological studies of rat brain slice cultures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium chloride, 1M aq. soln., sterile is used as an excellent magnesium ion source for synthesis and biology research. Magnesium chloride is widely used to supply the magnesium ion in various molecular biology applications, including PCR reactions and buffers. It was used to prepare buffer to lyse cells for immunoprecipitation1 and to prepare serum-free medium for electrophysiological studies of rat brain slice cultures.

Solubility
Soluble in water (542 mg/ml at 20°C), alcohol, methanol (15.5 g/100g at 0°C), and ethanol (3.61 g/100g at 0°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Very hygroscopic.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text