Potassium chloride, 1M aq. soln., RNAse free
Potassium chloride, 1M aq. soln., RNAse free
Potassium chloride, 1M aq. soln., RNAse free
Thermo Scientific Chemicals

Potassium chloride, 1M aq. soln., RNAse free

CAS: 7447-40-7 | ClK | 74.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J62422.AK
also known as J62422-AK
Price (EUR)
72,65
Online Exclusive
80,10
Save 7,45 (9%)
Each
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
72,65
Online Exclusive
80,10
Save 7,45 (9%)
Each
Specifications
CAS7447-40-7
ChEBICHEBI:32588
Chemical Name or MaterialPotassium chloride
Concentration or Composition (by Analyte or Components)1M aq. soln.
Formula Weight74.55
View more
It may be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins. This solution is used in a variety of biological applications, including DNA and RNA isolation techniques. Also used to establish a constant ionic strength in a sample solution. pH: 5.0-8.0 (25°C, 1 M in H2O)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It may be used for the preparation of phosphate buffered saline, and for the extraction and solubilization of proteins. This solution is used in a variety of biological applications, including DNA and RNA isolation techniques. Also used to establish a constant ionic strength in a sample solution. pH: 5.0-8.0 (25°C, 1 M in H2O)

Solubility
Not miscible in water.

Notes
Store at room temperature. Ensure good ventilation.
RUO – Research Use Only
Hideki Ando.; Toshiaki Furuta.; Hitoshi Okamoto. Photo-Mediated Gene Activation by Using Caged mRNA in Zebrafish Embryos.Methods in Cell Biology. 2004, 77,159-171 .