Melatonin, 99+%
Melatonin, 99+%
Melatonin, 99+%
Thermo Scientific Chemicals

Melatonin, 99+%

Melatonine, 99%, CAS # 73-31-4, is a hormone usually produced in the pineal gland and released into the blood. | CAS: 73-31-4 | C13H16N2O2 | 232.283 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J62452.14
also known as J62452-14
Price (USD)
162.65
Online exclusive
180.00
Save 17.35
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
162.65
Online exclusive
180.00
Save 17.35
Each
Chemical Identifiers
CAS73-31-4
IUPAC NameN-[2-(5-methoxy-1H-indol-3-yl)ethyl]acetamide
Molecular FormulaC13H16N2O2
InChI KeyDRLFMBDRBRZALE-UHFFFAOYSA-N
SMILESCOC1=CC=C2NC=C(CCNC(C)=O)C2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>99.0%
Melatonin acts as a neurohormone that upregulates antioxidant enzymes and inhibits rat NOS1. It also acts as a particularly significant endogenous antioxidant. Melatonin is a rat NOS1 (nNOS) inhibitor. Melatonin is an activator of MEL-1A-R and MEL-1B-R.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Melatonine is a hormone produced by the pineal gland that shows several biological activities, including the somnolence, sleep-wake cycle, circadian rhythm, stress response, aging, and immunity activities

• It shows antioxidant, autocoid, paracoid, hormonal, anti- inflammatory and anti-apoptotic activities

Applications

• Melatonin shows neurohormonal activity and can upregulate antioxidant enzymes and inhibit rat NOS1

• Melatonin has been reported to be an activator of MEL-1A-R and MEL-1B-R

• The production and secretion of melatonine are dependent on beta-adrenergic receptor function

• During melatonine synthesis, the essential amino acid L-tryptophan is used as a precursor

• This compound inhibits apoptosis in immune cells and neurons, and enhances apoptotic cell death of cancer cells

• It can inhibit proliferation and metastasis of breast cancer cells by modulating Rho-associated kinase protein-1 expression

RUO – Research Use Only

General References:

  1. Borin, T.F.; Arbab, A.S.; Gelaleti, G.B.; Ferreira, L.C.; Moschetta, M.G.; Jardim-Perassi, B.V.; Iskander, A.S.; Varma, N.R.; Shankar, A.; Coimbra, V.B.; Fabri, V.A.; de Oliveira, J.G.; Zuccari, D.A. Melatonin decreases breast cancer metastasis by modulating Rho-associated kinase protein-1 expression. J Pineal Res. 2016, 60, (1), 3-15.
  2. Tan, D.X.; Manchester, L.C.; Hardeland, R.; Lopez-Burillo, S.; Mayo, J.C.; Sainz, R.M.; Reiter, R.J. Melatonin: a hormone, a tissue factor, an autocoid, a paracoid, and an antioxidant vitamin. J Pineal Res. 2003, 34, (1), 75-78.
  3. Dubocovich, M.L. Pharmacology and function of melatonin receptors. FASEB J. 1988, 2, (12), 2765-73.
  4. Reiter, R.J., et al. Free radical-mediated molecular damage. Mechanisms for the protective actions of melatonin in the central nervous system. Ann. N.Y. Acad. Sci. 2001, 939, 200-15.