Tetraethylammonium chloride
Tetraethylammonium chloride
Tetraethylammonium chloride
Tetraethylammonium chloride
Thermo Scientific Chemicals

Tetraethylammonium chloride

Phase transfer catalyst | CAS: 56-34-8 | C8H20ClN | 165.71 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number J62507.14
also known as J62507-14
Price (EUR)
38,50
Each
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (EUR)
38,50
Each
Add to cart
Chemical Identifiers
CAS56-34-8
IUPAC Nametetraethylazanium chloride
Molecular FormulaC8H20ClN
InChI KeyYMBCJWGVCUEGHA-UHFFFAOYSA-M
SMILES[Cl-].CC[N+](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
FormCrystals or crystalline powder
Assay (unspecified)≥95.0%
Tetraethylammonium chloride is used as a catalyst. Tetraethylammonium Chloride can be used as a source of tetraethylammonium ions for various pharmaceutical studies and has the ability to block K+ channels in various tissues. Tetraethylammonium Chloride can also block the transmission of nervous impulses across autonomic ganglions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetraethylammonium chloride is used as a catalyst. Tetraethylammonium Chloride can be used as a source of tetraethylammonium ions for various pharmaceutical studies and has the ability to block K+ channels in various tissues. Tetraethylammonium Chloride can also block the transmission of nervous impulses across autonomic ganglions.

Solubility
Soluble in water, ethanol, chloroform, or acetone

Notes
Hygroscopic, Incompatible with strong oxidizing agents. Protect from moisture.  Keep container tightly closed in a dry and well-ventilated place. Handle under argon. Store in cool place.
RUO – Research Use Only

General References:

  1. Wei AD et al. Nomenclature and molecular relationships of calcium-activated potassium channels. Pharmacol Rev. 2005, 57, 463-72.
  2. Niu et al. Vasorelaxant effect of taurine is diminished by tetraethylammonium in rat isolated arteries. Eur.J.Pharmacol. 2008, 580, (1-2),169-74.