This product is animal origin and can only be shipped within the United States
RIPA buffer (5X)
Thermo Scientific Chemicals

RIPA buffer (5X)

RIPA buffer, radio immunoprecipitation assay buffer, is used for whole cell isolation of proteins from tissues and cell culture.
Have Questions?
Change viewbuttonViewtableView
Quantity:
50mL
100mL
Catalog number J62524.AD
also known as J62524-AD
Price (USD)
-
Quantity:
50mL
Request bulk or custom format
Specifications
Concentration5X
FormatLiquid
Quantity50mL
Product TypeCell Lysis Reagent
Unit SizeEach
RIPA buffer is used to lyse cells and tissues. It is used to lyse cultured mammalian cells from both plated cells and cells pelleted from suspension cultures. This buffer enables protein extraction from cytoplasmic, membrane and and is particularly useful for disruption of nuclear membranes in the preparation of nuclear extracts. It is compatible with many applications, including reporter assays, protein assays, immunoassays and protein purification. This product is ideal for general cell-tissue lysis and sub cellular organelle or molecular fractionation, including immunoprecipation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• RIPA can be used for isolation of cytoplasmic, membrane, nuclear, and mitochondrial proteins
• Protease inhibitors can be added to RIPA buffer to assist in protein isolation

Application

• RIPA buffer is suitable for use in protein isolation from culture cells or homogenized tissues
• Downstream applications of RIPA buffer include ELISA, western blot, immunoprecipitation, and SDS-PAGE

RUO – Research Use Only

General References:

  1. Mccoy, M.S.; Bargmann, C.I. Human colon carcinoma Ki-ras2 oncogene and its corresponding proto-oncogene. Mol. Cell. Biol. 1984, 4, (8), 1577-1582.
  2. Dorfman, T.; Bukovsky, A.; Ohagen, A.; Höglund, S. Functional domains of the capsid protein of human immunodeficiency virus type 1. J. Virol. 1994, 68, (12), 8180-8187.
Contents & Storage
2° to 8°C

Frequently asked questions (FAQs)

What are the concentrations of the components in RIPA buffer (5X) (Cat. No. J62524.xx)?

RIPA buffer (5X) contains 250 mM Tris-HCl (pH 7.4), 750 mM NaCl, 5% NP-40, 2.5% sodium deoxycholate, and 0.5% SDS.