BIS-TRIS, 0.2M buffer soln., pH 6.0
BIS-TRIS, 0.2M buffer soln., pH 6.0
BIS-TRIS, 0.2M buffer soln., pH 6.0
Thermo Scientific Chemicals

BIS-TRIS, 0.2M buffer soln., pH 6.0

CAS: 6976-37-0 | C8H19NO5 | 209.242 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J62686.AK
also known as J62686-AK
Price (EUR)
110,65
Online Exclusive
123,00
Save 12,35 (10%)
Each
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
110,65
Online Exclusive
123,00
Save 12,35 (10%)
Each
Specifications
CAS6976-37-0
ChEBICHEBI:41250
Chemical Name or MaterialBIS-TRIS
ColorColorless
IUPAC Name2-[bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only