Thermo Scientific Chemicals

Orange G, Electrophoresis Grade, Thermo Scientific Chemicals

Catalog number: J62743.A1
1 kg, Each
Thermo Scientific Chemicals

Orange G, Electrophoresis Grade, Thermo Scientific Chemicals

Catalog number: J62743.A1
1 kg, Each
Quantity
Catalog number: J62743.A1
also known as J62743-A1
Price (USD)
Price: 179.00
Online price: 152.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1936-15-8
IUPAC Name
disodium (8Z)-7-oxo-8-(2-phenylhydrazin-1-ylidene)-7,8-dihydronaphthalene-1,3-disulfonate
Molecular Formula
C16H10N2Na2O7S2
InChI Key
AEOVEGJBKQQFOP-DDVLFWKVSA-L
SMILES
[Na+].[Na+].[O-]S(=O)(=O)C1=CC2=C(\C(=N\NC3=CC=CC=C3)C(=O)C=C2)C(=C1)S([O-])(=O)=O
Loss on Drying
10.0% max.
Assay (unspecified)
Dye content: 90% min.
Assay (unspecified)
Lambda max: 479nm

Description

Collagen stain for connective tissue

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text