Thermo Scientific Chemicals

Benzamidine hydrochloride hydrate, 99%, water ca 10-14%, Thermo Scientific Chemicals

Catalog number: J62823.09
10 g, Each
Thermo Scientific Chemicals

Benzamidine hydrochloride hydrate, 99%, water ca 10-14%, Thermo Scientific Chemicals

Catalog number: J62823.09
10 g, Each
Quantity
Catalog number: J62823.09
also known as J62823-09
Price (USD)
Price: 61.40
Online price: 53.65
Your price:
Quantity
-

Chemical Identifiers

CAS
206752-36-5
IUPAC Name
benzenecarboximidamide hydrochloride
Molecular Formula
C7H9ClN2
InChI Key
LZCZIHQBSCVGRD-UHFFFAOYSA-N
SMILES
Cl.NC(=N)C1=CC=CC=C1
Assay from Supplier's CofA
≥98.5%
Appearance (Color)
White
Form
Powder
Water Content (Karl Fischer Titration)
10.0-14.0%

Description

Reversible inhibitor of trypsin, trypsin-like proteins and serine proteases

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text