BIS-TRIS propane, 0.2M buffer soln., pH 8.0
BIS-TRIS propane, 0.2M buffer soln., pH 8.0
BIS-TRIS propane, 0.2M buffer soln., pH 8.0
Thermo Scientific Chemicals

BIS-TRIS propane, 0.2M buffer soln., pH 8.0

CAS: 64431-96-5 | C11H26N2O6 | 282.337 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J62831.AK
also known as J62831-AK
Price (EUR)
112,00
Each
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
112,00
Each
Specifications
CAS64431-96-5
ChEBICHEBI:40947
Chemical Name or MaterialBIS-TRIS propane buffer soln.
IUPAC Name2-[3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propylamino]-2-(hydroxymethyl)propane-1,3-diol
InChI KeyHHKZCCWKTZRCCL-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only