Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane hydrochloride, 1M soln., pH 7,4, RNase free

Catalog number: J62848.AK
250 mL, Each
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane hydrochloride, 1M soln., pH 7,4, RNase free

Catalog number: J62848.AK
250 mL, Each
Quantity
Catalog number: J62848.AK
also known as J62848-AK
Price (USD)
Price: 80.20
Your Price:
Quantity
-

Chemical Identifiers

CAS
1185-53-1
IUPAC Name
hydrogen 2-amino-2-(hydroxymethyl)propane-1,3-diol chloride
Molecular Formula
C4H12ClNO3
InChI Key
QKNYBSVHEMOAJP-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NC(CO)(CO)CO
pH
7.4 ± 0.15
RNase
None detected
Appearance (Color)
Clear colorless to light cream
Form
Liquid
DNase
None detected

Description

Tris Buffered Saline (TBS) is a pH stabilizing solution used for western blot and ELISA procedures. It is used as a convenient prepared buffer for use in various applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text