Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific Chemicals

Protamine sulfate, Thermo Scientific Chemicals

Catalog number: J62926.06
5 g, Each
Thermo Scientific Chemicals

Protamine sulfate, Thermo Scientific Chemicals

Catalog number: J62926.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J62926.06
also known as J62926-06
Price (USD)
Price: 125.00
Your price:
Quantity
-

Chemical Identifiers

CAS
9009-65-8
SMILES
*
Molecular Weight (g/mol)
0.00
Loss on Drying
5.0% max.
Appearance (Color)
White to off-white
Assay (unspecified)
Each mg neutralizes not less than 100 USP Heparin units (dry basis)
Form
Powder

Description

It is suitable for personal care, cosmetic and pharmaceutical applications. It is used widely to reverse the anticoagulant effects of heparin. It acts as a heparin antagonist. It is also a weak anticoagulant. It may be used for the removal of DNA from protein samples or the purification of DNA binding proteins. Protamine sulfate may also be used as an alternative to polybrene in retroviral mediated gene transfer. Protamine sulfate has antibacterial properties and inhibits pepsin activity. Protamine sulfate and other polycations have been shown to inhibit the classical pathway of complement. It neutralizes the anticoagulant activity of heparin, and inhibits turnover of lipoproteins by lipoprotein lipase. The aggregation of erythrocytes in the presence of protamine sulfate, for application to the prediction of MN phenotypes, has been investigated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
It is sparingly soluble in water.

Notes
Hygroscopic. Store at room temperature. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text