HEPES, 0.5M buffer soln., pH 8.0
HEPES, 0.5M buffer soln., pH 8.0
HEPES, 0.5M buffer soln., pH 8.0
Thermo Scientific Chemicals

HEPES, 0.5M buffer soln., pH 8.0

CAS: 7365-45-9 | C8H18N2O4S | 238.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J63002.AE
also known as J63002-AE
Price (EUR)
66,65
Online Exclusive
73,40
Save 6,75 (9%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
66,65
Online Exclusive
73,40
Save 6,75 (9%)
Each
Specifications
CAS7365-45-9
ChEBICHEBI:42334
Chemical Name or MaterialHEPES buffer soln.
IUPAC Name2-[4-(2-hydroxyethyl)piperazin-1-yl]ethanesulfonic acid
InChI KeyJKMHFZQWWAIEOD-UHFFFAOYSA-N
View more

HEPES has been utilized extensively for research and commercial applications in the cell culture and tissue culture industries. It is useful in the pH range 6.8-8.2. HEPES is used as a zwitterionic, all-purpose Good’s buffer for use in biological research. HEPES is widely used in cell culture, largely because it is better at maintaining physiological pH despite changes in carbon dioxide concentration when compared to bicarbonate buffers, which are also commonly used in cell culture.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES has been utilized extensively for research and commercial applications in the cell culture and tissue culture industries. It is useful in the pH range 6.8-8.2. HEPES is used as a zwitterionic, all-purpose “Good’s” buffer for use in biological research. HEPES is widely used in cell culture, largely because it is better at maintaining physiological pH despite changes in carbon dioxide concentration when compared to bicarbonate buffers, which are also commonly used in cell culture.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
David M.L. Goodgame.; Martin Joy, A. EPR study of the Cr(V) and radical species produced in the reduction of Cr(VI) by ascorbate. Inorganica Chimica Acta. 1987, 135, (2), 115-118.