Neutral Red
Neutral Red
Neutral Red
Thermo Scientific Chemicals

Neutral Red

Nuclear counterstain useful in pH range 6.8 to 8.0 | CAS: 553-24-2 | C15H17ClN4 | 288.779 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number J63054.06
also known as J63054-06
Price (EUR)
59,65
Online Exclusive
66,30
Save 6,65 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (EUR)
59,65
Online Exclusive
66,30
Save 6,65 (10%)
Each
Chemical Identifiers
CAS553-24-2
IUPAC Namehydrogen N2,N2,7-trimethylphenazine-2,8-diamine chloride
Molecular FormulaC15H17ClN4
InChI KeyPGSADBUBUOPOJS-UHFFFAOYSA-N
SMILES[H+].[Cl-].CN(C)C1=CC=C2N=C3C=C(C)C(N)=CC3=NC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Solution TestSolubility 0.1% (50% ethanol and 2 ml acetic acid): Clear red solution
Appearance (Color)Very dark green
Assay (special)Dye content: ≥ 50%
FormPowder
Nuclear counterstain useful in pH range 6.8 - 8.0

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only