Thermo Scientific™

2'-Deoxycytidine

25g, Each
Thermo Scientific™

2'-Deoxycytidine

25g, Each
Quantity
Catalog number: J63062.14
also known as J63062-14
Price (USD)
Price: 335.00 / Each
Your Price:
Quantity
-

Chemical Identifiers

CAS
951-77-9
IUPAC Name
4-amino-1-[4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2-dihydropyrimidin-2-one
Molecular Formula
C9H13N3O4
InChI Key
CKTSBUTUHBMZGZ-UHFFFAOYNA-N
SMILES
NC1=NC(=O)N(C=C1)C1CC(O)C(CO)O1
Appearance (Color)
White to off-white
Form
Powder
Assay (HPLC)
97.0% min

Description

2?-Deoxycytidine (deoxyC) is one of the deoxynucleosides which after phosphorylation to dCTP is used to synthesis DNA via various DNA polymerases or reverse transcriptases. 2?-Deoxycytidine (deoxyC) is the substrate for deoxycytidine deaminase (EC 3.5.4.14) which converts it into 2?-deoxyuridine. 2?-Deoxycytidine is phosphorylated to the nucleotide dCMP by the enzyme deoxycytidine kinase (DCK).

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2′-Deoxycytidine (deoxyC) is one of the deoxynucleosides which after phosphorylation to dCTP is used to synthesis DNA via various DNA polymerases or reverse transcriptases. 2′-Deoxycytidine (deoxyC) is the substrate for deoxycytidine deaminase (EC 3.5.4.14) which converts it into 2′-deoxyuridine. 2′-Deoxycytidine is phosphorylated to the nucleotide dCMP by the enzyme deoxycytidine kinase (DCK).

Solubility
Soluble in water, DMSO.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.
RUO – Research Use Only

Figures