Ponceau S, 0.1% v/v soln. in 5% acetic acid
Ponceau S, 0.1% v/v soln. in 5% acetic acid
Ponceau S, 0.1% v/v soln. in 5% acetic acid
Thermo Scientific Chemicals

Ponceau S, 0.1% v/v soln. in 5% acetic acid

CAS: 6226-79-5 | C22H12N4Na4O13S4 | 760.552 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number J63139.K2
also known as J63139-K2
Price (USD)
179.00
Each
Quantity:
1 L
Request bulk or custom format
Price (USD)
179.00
Each
Specifications
CAS6226-79-5
Chemical Name or MaterialPonceau S
ColorRed
Concentration0.2% v/v soln. in 5% acetic acid
Formula Weight760.56
View more
Ponceau S is used to prepare a stain for rapid reversible detection of protein bands on nitrocellulose or polyvinylidene fluoride membranes, as well as on cellulose acetate membranes. And it is also used as a rapid protein band stain.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only