Manganese(II) chloride, 1M aq. soln.
Manganese(II) chloride, 1M aq. soln.
Manganese(II) chloride, 1M aq. soln.
Thermo Scientific Chemicals

Manganese(II) chloride, 1M aq. soln.

CAS: 7773-01-5 | Cl2Mn | 125.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
100 mL
Catalog number J63150.AE
also known as J63150-AE
Price (EUR)
76,65
Online Exclusive
84,90
Save 8,25 (10%)
Each
Quantity:
100 mL
Request bulk or custom format
Price (EUR)
76,65
Online Exclusive
84,90
Save 8,25 (10%)
Each
Specifications
CAS7773-01-5
ChEBICHEBI:63041
Chemical Name or MaterialManganese(II) chloride
ColorColorless
Concentration or Composition (by Analyte or Components)1M aq. soln.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only