Thermo Scientific Chemicals

Cyclosporin A, 99+%, Thermo Scientific Chemicals

Catalog number: J63191.03
1 g, Each
Thermo Scientific Chemicals

Cyclosporin A, 99+%, Thermo Scientific Chemicals

Catalog number: J63191.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63191.03
also known as J63191-03
Price (USD)
Quantity
-

Chemical Identifiers

CAS
59865-13-3
Appearance (Color)
White
Assay from Suppliers CofA
≥99.0%
Form
Powder or crystals or crystalline powder

Description

Ciclosporin A induces apoptosis and inhibits angiogenesis induced by VEGF. It also suppresses the replication of hepatitis C virus genome in cultured hepatocytes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• Cyclosporin A is a non-polar cyclic oligopeptide produced by the fungus Tolypocladium inflatum
Applications
• It inhibits phosphoprotein phosphatase
• Cyclosporin A binds to the cellular protein cytophilin and inhibits the enzyme calcineurin
• Furthermore, it can induce apoptosis and inhibit angiogenesis induced by VEGF
• It exerts potent immunosuppressive activity on T immune cells, and causes reversible inhibition of immunocompetent lymphocytes in the G0- and G1-phase of the cell cycle
• It also suppresses the hepatitis C virus replication in cultured hepatocytes
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text