4,5',8-Trimethylpsoralen, 99%
4,5',8-Trimethylpsoralen, 99%
4,5',8-Trimethylpsoralen, 99%
4,5',8-Trimethylpsoralen, 99%
Thermo Scientific Chemicals

4,5',8-Trimethylpsoralen, 99%

CAS: 3902-71-4 | C14H12O3 | 228.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63226.06
also known as J63226-06
Price (EUR)
343,00
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (EUR)
343,00
Each
Add to cart
Specifications
Beilstein221723
CAS3902-71-4
ChEBICHEBI:28329
Chemical Name or Material4,5',8-Trimethylpsoralen
ColorWhite
View more
4,5',8-Trimethylpsoralen is used for treatment of psoriasis, and as a photochemical crosslinker of DNA used as a probe for nucleic acid structure, used to crosslink DNA onto mica surfaces, mutagenesis of N2 worms.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,5′,8-Trimethylpsoralen is used for treatment of psoriasis, and as a photochemical crosslinker of DNA used as a probe for nucleic acid structure, used to crosslink DNA onto mica surfaces, mutagenesis of N2 worms.

Solubility
Soluble in chloroform

Notes
Keep container tightly closed. Store away from oxidizing agents. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Aba Losi.; Roberta Bedotti.; Cristiano Viappiani. Time-Resolved Photoacoustics Determination of Intersystem Crossing and Singlet Oxygen Photosensitization Quantum Yields for 4,5',8-Trimethylpsoralen.J. Phys. Chem.1995, 99, (43), 16162-16167.
  2. Kandallu R. Kumaresan.; Muthukumar Ramaswamy.; Anthony T. Yeung. Structure of the DNA interstrand crosslink of 4,5',8-trimethylpsoralen.Biochemistry.1992, 31, (29), 6774-6783.