Thermo Scientific Chemicals
Indomethacin, 99+%, Thermo Scientific Chemicals
Catalog number: J63255.14
25 g, Each
Thermo Scientific Chemicals
Indomethacin, 99+%, Thermo Scientific Chemicals
Catalog number: J63255.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63255.14
also known as J63255-14
Price (USD)
Price: 203.00
Online price: 172.65
Your price:
Quantity
-
Description
Non-selective cyclooxygenase inhibitorIndomethacin acts as an antiinflammatory, antipyretic and analgesic. It serves as an inhibitor cyclooxygenase such as Cox-1 and Cox-2. In addition to this, it is used as a prescription medication in order to reduce fever, stiffness, swelling and pain.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Indomethacin is a synthetic NSAID of the indole family that inhibits cyclooxygenase and reduces prostaglandin and pyrogen release
• Indomethacin may suppress the multi-drug resistant transporter and thereby increase the efficacy of antineoplastics
• Indomethacin activates phosphatases that inhibit signaling pathways associated with cancer cell migration and proliferation
Applications
• Indomethacin may be used for the laboratory study of cancer cell proliferation, tumorigenesis and drug interactions
• Indomethacin may be used for laboratory study of the effect of inhibiting the multi-drug resistance transporter, or for downregulation of survival signaling in cells via survivin