MES, 0.2M buffer soln., pH 5.5
MES, 0.2M buffer soln., pH 5.5
MES, 0.2M buffer soln., pH 5.5
Thermo Scientific Chemicals

MES, 0.2M buffer soln., pH 5.5

CAS: 145224-94-8 | C6H15NO5S | 213.248 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
250 mL
500 mL
產品號碼 J63257.AK
亦稱為 J63257-AK
價格 (TWD)
-
Quantity:
250 mL
Request bulk or custom format
規格
CAS145224-94-8
Chemical Name or MaterialMES buffer soln.
ColorColorless
Health Hazard 1Warning
Health Hazard 2GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
檢視更多
MES is used as a running buffer for resolving very small proteins on bis-tris gels, also used as a biological buffer in plant cell cultures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES is used as a running buffer for resolving very small proteins on bis-tris gels, also used as a biological buffer in plant cell cultures.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Leonga, K.W.; Maoa, H,Q.; Truong-Leb, V.L.; Roya, K.; Walsha, S.M.; August, J.T. DNA-polycation nanospheres as non-viral gene delivery vehicles.Journal of Controlled Release.1998, 53, (1-3), 183-193.