BioAcryl-P (40%, 39:1)
Thermo Scientific Chemicals

BioAcryl-P (40%, 39:1)

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
Catalog number J63323.K2
also known as J63323-K2
Price (TWD)
-
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialAcrylamide/Bisacrylamide 39:1
Concentration40% soln
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ACRYLAMIDE SOLUTION
Health Hazard 1H302+H332-H315-H317-H319-H340-H350-H361f-H372
Health Hazard 2GHS H Statement
H340-H350-H372-H361-H302-H315-H319-H317
May cause genetic defects.
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
View more
Acrylamide is used for sequencing gels and separation of proteins, used in protein and nucleic acid electrophoresis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acrylamide is used for sequencing gels and separation of proteins, used in protein and nucleic acid electrophoresis.

Solubility
Fully miscible in water.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Daekyu Sun.; Laurence H. Hurley. Biochemical Techniques for the Characterization of G-Quadruplex Structures: EMSA, DMS Footprinting, and DNA Polymerase Stop Assay.G-Quadruplex DNA. 2009, 608, 65-79.
  2. Stolnika, S.; Daudalia, B.; Ariena, A.; Whetstonea, J.; Healda, C.R.; Garnetta, M.C.; Davisa, S.; Illum, L. The effect of surface coverage and conformation of poly(ethylene oxide) (PEO) chains of poloxamer 407 on the biological fate of model colloidal drug carriers.Biochimica et Biophysica Acta (BBA) - Biomembranes.2001, 1514, (2), 261-279.