Informational:Informational:This product is animal origin and can only be shipped within the United States
Thermo Scientific™
RIPA buffer (2X)
125mL
Thermo Scientific™
RIPA buffer (2X)
125mL
Quantity
Catalog number: J63324.EQE
also known as J63324-=E
Price (USD)
Description
RIPA is used as a high quality, ready to use mammalian cell and tissue lysis buffer. If protease inhibitors are included after receipt, it can be used to lyse cells and tissue, for radio immunoprecipitation assay.
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
RUO – Research Use Only