Thermo Scientific Chemicals

MES, 1.0M buffer soln., pH 5.5

Catalog number: J63341.AP
500 mL, Each
Thermo Scientific Chemicals

MES, 1.0M buffer soln., pH 5.5

Catalog number: J63341.AP
500 mL, Each
Quantity
Catalog number: J63341.AP
also known as J63341-AP
Price (USD)
Price: 238.00
Online price: 202.65
Your price:
Quantity
-

Chemical Identifiers

CAS
145224-94-8
IUPAC Name
4-(2-sulfonatoethyl)morpholin-4-ium
Molecular Formula
C6H13NO4S
InChI Key
SXGZJKUKBWWHRA-UHFFFAOYSA-N
SMILES
[O-]S(=O)(=O)CC[NH+]1CCOCC1
Appearance (Color)
Clear pale yellow
Comment
Component is dissolved in UV treated 18.2 megohm-cm ± 1 water, followed by pH adjustment and filtered through sub-micron filter.
pH
5.5 ± 0.1 (after dilution is water)
Form
Liquid

Description

MES is used as a buffering agent in biology and biochemistry. It is utilized for resolving very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MES is used as a buffering agent in biology and biochemistry. It is utilized for resolving very small proteins on bis-tris gels. It is mainly used as a biological buffer in plant cell cultures. Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text