Potassium acetate, 8M aq. soln.
Potassium acetate, 8M aq. soln.
Potassium acetate, 8M aq. soln.
Potassium acetate, 8M aq. soln.
Thermo Scientific Chemicals

Potassium acetate, 8M aq. soln.

CAS: 127-08-2 | C2H3KO2 | 98.142 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
250 mL
Catalog number J63372.AK
also known as J63372-AK
Price (EUR)
68,00
Online Exclusive
80,00
Save 12,00 (15%)
Each
Add to cart
Quantity:
250 mL
Request bulk or custom format
Price (EUR)
68,00
Online Exclusive
80,00
Save 12,00 (15%)
Each
Add to cart
Specifications
CAS127-08-2
ChEBICHEBI:32029
Chemical Name or MaterialPotassium acetate
Concentration8M aq. soln.
Formula Weight98.15
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only