Sodium n-dodecyl sulfate (SDS), 20% aq. soln.
Sodium n-dodecyl sulfate (SDS), 20% aq. soln.
Sodium n-dodecyl sulfate (SDS), 20% aq. soln.
Thermo Scientific Chemicals

Sodium n-dodecyl sulfate (SDS), 20% aq. soln.

CAS: 151-21-3 | C12H25NaO4S | 288.38 g/mol
Have Questions?
產品號碼Quantity
J63394.AP
亦稱為 J63394-AP
500 mL
產品號碼 J63394.AP
亦稱為 J63394-AP
價格 (TWD)
-
Quantity:
500 mL
Request bulk or custom format
化學識別
CAS151-21-3
IUPAC Namesodium dodecyl sulfate
Molecular FormulaC12H25NaO4S
InChI KeyDBMJMQXJHONAFJ-UHFFFAOYSA-M
SMILES[Na+].CCCCCCCCCCCCOS([O-])(=O)=O
檢視更多
規格Specification Sheet規格表
FormLiquid
Appearance (Color)Colorless
CommentProduct may precipitate at low temperatures. Heat at 40°C for 15 minutes to redissolve.

Sodium n-dodecyl sulfate 20% aq. solution is used as an anionic surfactant. It is also used as fat emulsifier, a wetting agent, a detergent in cosmetics, pharmaceuticals and toothpastes. It also serves as a research tool in protein biochemistry. It is mainly used in detergents for laundry with many cleaning applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium n-dodecyl sulfate 20% aq. solution is used as an anionic surfactant. It is also used as fat emulsifier, a wetting agent, a detergent in cosmetics, pharmaceuticals and toothpastes. It also serves as a research tool in protein biochemistry. It is mainly used in detergents for laundry with many cleaning applications.

Solubility
Miscible with ethanol.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Zhao, M.; Yuan, J.; Zheng, L. Spontaneous formation of vesicles by N-dodecyl-N-methylpyrrolidinium bromide (C 12 MPB) ionic liquid and sodium dodecyl sulfate (SDS) in aqueous solution. Colloids Surf., A 2012, 407, 116-120.
  2. Chen, S.; Duhamel, J.; Peng, B.; Zaman, M.; Tam, K. C. Interactions between a Series of Pyrene End-Labeled Poly (ethylene oxide) s and Sodium Dodecyl Sulfate in Aqueous Solution Probed by Fluorescence. Langmuir 2014, 30, (44), 13164-13175.