Thermo Scientific Chemicals

Imidazole, 0.5M buffer soln., pH 7.5

Catalog number: J63496.AK
250 mL, Each
Thermo Scientific Chemicals

Imidazole, 0.5M buffer soln., pH 7.5

Catalog number: J63496.AK
250 mL, Each
Quantity
Catalog number: J63496.AK
also known as J63496-AK
Price (USD)
Price: 151.00
Online price: 128.65
Your price:
Quantity
-

Description

Imidazole is used as a chelator for the binding of various divalent cations, and is also used to elute tagged proteins bound to Ni ions attached to the surface of beads in the chromatography column.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Imidazole is used as a chelator for the binding of various divalent cations, and is also used to elute tagged proteins bound to Ni ions attached to the surface of beads in the chromatography column.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text