Thermo Scientific Chemicals

L-Tyrosine, Cell Culture Reagent

Catalog number: J63511.09
10 g, Each
Thermo Scientific Chemicals

L-Tyrosine, Cell Culture Reagent

Catalog number: J63511.09
10 g, Each
Quantity
Catalog number: J63511.09
also known as J63511-09
Price (USD)
Price: 21.20
Online price: 17.65
Your price:
Quantity
-

Chemical Identifiers

CAS
60-18-4
IUPAC Name
(2S)-2-amino-3-(4-hydroxyphenyl)propanoic acid
Molecular Formula
C9H11NO3
InChI Key
OUYCCCASQSFEME-SVGMAFHSNA-N
SMILES
N[C@@H](CC1=CC=C(O)C=C1)C(O)=O
Loss on Drying
≤0.5%
Appearance (Color)
White
Assay from Supplier's CofA
≥98.5%
Heavy Metals
≤15 ppm
Impurity content
Iron: ≤30 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text