Bombesin
Thermo Scientific Chemicals

Bombesin

For cellular and molecular biology applications
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 mg
5 mg
Catalog number J63514.MCR
also known as J63514-M^
Price (EUR)
55,80
Each
Quantity:
1 mg
Request bulk or custom format
Price (EUR)
55,80
Each
Specifications
For Use With (Application)Gut tetradecapeptide with the ability to stimulate release of various hormones
FormatPowder
Molecular Weight (g/mol)1619.90
Product TypeBombesin
Purification MethodHPLC
View more
Gut tetradecapeptide with the ability to stimulate release of various hormones

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Contents & Storage
2°C to 8°C