Thermo Scientific Chemicals

Domperidone, Thermo Scientific Chemicals

Catalog number: J63681.ME
500 mg, Each
Thermo Scientific Chemicals

Domperidone, Thermo Scientific Chemicals

Catalog number: J63681.ME
500 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63681.ME
also known as J63681-ME
Price (USD)
Quantity
-

Description

Domperidone is used as a Peripheral dopamine receptor antagonist that does not cross the blood-brain barrier. The antidopaminergic effects of Domperidone have been correlated to antiemetic and gastroprokinetic action.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Domperidone is used as a Peripheral dopamine receptor antagonist that does not cross the blood-brain barrier. The antidopaminergic effects of Domperidone have been correlated to antiemetic and gastroprokinetic action.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text