Thermo Scientific Chemicals

Clonidine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: J63707.03
1 g, Each
Thermo Scientific Chemicals

Clonidine hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: J63707.03
1 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63707.03
also known as J63707-03
Price (USD)
Quantity
-

Chemical Identifiers

CAS
4205-91-8
IUPAC Name
hydrogen N-(2,6-dichlorophenyl)-4,5-dihydro-1H-imidazol-2-amine chloride
Molecular Formula
C9H10Cl3N3
InChI Key
ZNIFSRGNXRYGHF-UHFFFAOYSA-N
SMILES
[H+].[Cl-].ClC1=CC=CC(Cl)=C1NC1=NCCN1
Assay (HPLC)
>98.0%

Description

Clonidine hydrochloride tablets are indicated in the treatment of hypertension. It has found new uses, including treatment of some types of neuropathic pain, opioid detoxification, sleep hyperhidrosis, anaesthetic use, and off-label, to counter the side effects of stimulant medications such as methylphenidate or amphetamine. It is becoming a more accepted treatment for insomnia, as well as for relief of menopausal symptoms. Clonidine(4205-91-8) is increasingly used in conjunction with stimulants to treat attention-deficit hyperactivity disorder (ADHD).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water (50 mg/ml), DMSO (75 mM), methanol, chloroform (slightly), and dehydrated alcohol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text