L-Cystine dihydrochloride, 98%
L-Cystine dihydrochloride, 98%
L-Cystine dihydrochloride, 98%
Thermo Scientific Chemicals

L-Cystine dihydrochloride, 98%

CAS: 30925-07-6 | C6H14Cl2N2O4S2 | 313.208 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Catalog number J63717.30
also known as J63717-30
Price (EUR)
153,65
Online Exclusive
170,00
Save 16,35 (10%)
Each
Quantity:
250 g
Request bulk or custom format
Price (EUR)
153,65
Online Exclusive
170,00
Save 16,35 (10%)
Each
Chemical Identifiers
CAS30925-07-6
IUPAC Namedihydrogen (2R)-2-amino-3-[(2-amino-2-carboxyethyl)disulfanyl]propanoic acid dichloride
Molecular FormulaC6H14Cl2N2O4S2
InChI KeyHHGZUQPEIHGQST-VBCXBIGUSA-N
SMILES[H+].[H+].[Cl-].[Cl-].NC(CSSC[C@H](N)C(O)=O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Titration)≥97.5% (dried basis)
Residue on ignition<0.10%
Appearance (Color)White to pale yellow
Heavy Metals(as Pb): ≤20ppm
Loss on Drying≤1.0%
View more
Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement.

Solubility
Freely soluble in Dilute hydrochloric acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

General References:

  1. Yukio Kurita.; Walter Gordy. Electron Spin Resonance in a Gamma-Irradiated Single Crystal of L-Cystine Dihydrochloride.J. Chem. Phys. 1961, 34 (1),282 .
  2. Kazuyuki Akasaka.; Shun-ichi Ohnishi.; Tokuo Suita. ;Isamu Nitta. ESR Study of a Single Crystal of L-Cystine Dihydrochloride Irradiated at Low Temperature.J. Chem. Phys. 1964, 40, (40),3110 .