Thermo Scientific Chemicals

L-Cystine dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: J63717.A1
1 kg, Each
Thermo Scientific Chemicals

L-Cystine dihydrochloride, 98%, Thermo Scientific Chemicals

Catalog number: J63717.A1
1 kg, Each
Quantity
Catalog number: J63717.A1
also known as J63717-A1
Price (USD)
Price: 689.00
Online price: 585.65
Your price:
Quantity
-

Chemical Identifiers

CAS
30925-07-6
IUPAC Name
dihydrogen (2R)-2-amino-3-[(2-amino-2-carboxyethyl)disulfanyl]propanoic acid dichloride
Molecular Formula
C6H14Cl2N2O4S2
InChI Key
HHGZUQPEIHGQST-VBCXBIGUSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].NC(CSSC[C@H](N)C(O)=O)C(O)=O
Form
Crystalline powder
Chloride content
22.07-23.50%
Infrared spectrum
matches reference
Assay (Titration)
≥97.5% (dried basis)
Residue on ignition
<0.10%

Description

Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement.

Solubility
Freely soluble in Dilute hydrochloric acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text